missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivapure™ miniprep A / G spin columns

Product Code. 11725056 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
16 Pack
2 Pack
48 Pack
Each
Type:
Miniprep Spin Column
Spin Column
Matrix:
Protein A Resin
Protein G Resin
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Type Matrix unitSize
11725056 Each Miniprep Spin Column Protein G Resin Each
10246694 Each Miniprep Spin Column Protein A Resin Each
11715056 48 Pack Spin Column Protein A Resin Each
11724059 16 Pack Spin Column Protein A Resin Each
11750845 48 Pack Spin Column Protein G Resin Each
11760845 16 Pack Spin Column Protein G Resin Each
11770845 2 Pack Miniprep Spin Column Protein A Resin Each
11780845 2 Pack Miniprep Spin Column Protein G Resin Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 11725056 Supplier Sartorius Supplier No. VSARGBUFPK

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Centrifugal units with Protein A or Protein G resin for fast affinity purification of antibodies from small samples up to 0.65 ml.

Vivapure miniprep and spin columns are pre-packed centrifugal devices for purification of monoclonal and polyclonal antibodies from serum, ascites and cell culture supernatants. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many samples are processed in parallel. Vivapure miniprep & spin columns can conveniently be used in a centrifuge with swing-out rotor for standard 2.2 ml tubes. Binding capacity is about 1 mg IgG per unit. The low hold-up volume ensures high solute recovery with minimal loss by non-specific adsorption. Vivapure miniprep and spin columns can be re-used three times and are available as complete kits including buffers or as bulk packs.They are an attractive alternative to lengthy chromatographic methods such as FPLC. For larger volumes, the Vivapure Maxiprep kit is also availablity.

  • Spin column based kit
  • Devices are ready to use
  • Rapid purification with high yields
  • Parallel processing
  • Cost effective through re-use option
  • Starter kits available

TRUSTED_SUSTAINABILITY

Specifications

Matrix Protein G Resin
Quantity Each
For Use With (Application) Purification of Monoclonal and Polyclonal Antibodies from Serum, ascites and cell culture supernatants
Type Miniprep Spin Column
Sufficient For 4 Maxi or 48 miniprep Protein A and G spin column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.