Learn More
Sartorius Vivapure™ Mini S&Q H Starter Kit
Product Code. 11780855 Shop All Sartorius Products
Click to view available options
:
Vivapure Mini S and Q H; Binding capacity 1 to 4mg
Product Code. 11780855
Brand: Sartorius VSIX01SQ16
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ready-to-use centrifugal devices
Vivapure Mini Ion Exchange spin columns are centrifugal devices, based on Sartobind Membrane Adsorber technology as their chromatography matrix. The devices are ready-to-use and cannot run dry by their design. For many protein purification applications, they can replace time-consuming column chromatography. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many samples are processed parallel. Vivapure Mini is available with three different membrane chemistries and with medium (1mg/unit) or high (4mg/unit) protein binding capacity. Vivapure Mini units are intended for use in swing-out rotors and accommodate 0.5mL and 0.4mL samples, respectively.
  • Fast centrifugal spin columns
  • Easy and ready-to-use
  • Binding capacity 1mg up to 4mg protein
  • Ideal for parallel processing
TRUSTED_SUSTAINABILITY
Specifications
Bed Volume 240 μL
Capacity (Metric) 0.4 mL
Material Polypropylene
Pore Size 3 to 5 μm
Support Type Stabilized regenerated Cellulose
Quantity 16 Pack
For Use With (Application) Sample fractionation, purification condition scouting, small scale purification
Type Spin Column