missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivapure™ maxiprep A / G spin columns

Product Code. 11795046 Shop All Sartorius Products
Click to view available options
Quantity:
12 Pack
4 Pack
Support Type:
Protein A Resin
Protein G Resin
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11795046

Brand: Sartorius VSARAMAXIB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Centrifugal units with Protein A or Protein G resin for fast affinity purification of antibodies from samples up to and larger than 20 ml

Vivapure maxiprep Spin columns are pre-packed centrifugal devices for purification of monoclonal and polyclonal antibodies from serum, ascites and cell culture supernatants. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many samples are processed in parallel. Vivapure maxiprep Spin columns can conveniently be used in a centrifuge with swing-out rotor for standard 50 ml tubes. Binding capacity is about 20 mg IgG per unit. The low hold-up volume ensures high solute recovery with minimal loss by non-specific adsorption. Vivapure maxiprep Spin columns can be re-used five times and are available as complete kits including buffers or as bulk packs. They are an attractive alternative to lengthy chromatographic methods such as FPLC. For processing larger sample volumes than 20 ml, e. g. from diluted cell culture supernatants, the Vivapure maxiprep Spin columns can additionally be used with a sealing cap and pump collar for peristaltic pumps. For smaller volumes, the Vivapure Miniprep kit is also availablity.

  • Spin column based kit
  • Devices are ready to use
  • Rapid purification with high yields
  • Parallel processing
  • Cost effective through re-use option
  • Starter kits available
  • Processing of larger samples with peristaltic pump

TRUSTED_SUSTAINABILITY

Specifications

Support Type Protein A Resin
Quantity 12 Pack
For Use With (Application) Purification of Monoclonal and Polyclonal Antibodies from Serum, ascites and cell culture supernatants
Type Spin Column
Sufficient For 5 Users
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.