missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Economic and user-friendly
- No need for additional equipment, pressure or a vacuum source
- Individual devices with disposable plastic stands for up to four units
- Simple concentration technique:
- Just fill the unit with the solution to be concentrated
- Wait for the desired concentration level to be achieved
- Then pipet the concentrated sample from the dead-stop reservoir
- Separate concentration chambers eliminate the risk of cross-contamination
- Each unit features a high-flow, 7.5 kDa PES membrane
- Ideal for samples that are sensitive to shearing forces or to pressure-induced stress
- Well suited for samples that require processing in a cold room environment
- Perfect for a variety of clinical and general research applications
- Urine concentration and subsequent analysis of Bence-Jones proteins by gel electrophoresis
- Concentrating macromolecules in cerebrospinal fluid (CSF) or other biological solutions
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Ultrafiltration |
| Material (Membrane) | PES |
| MWCO | 7.5 kDa |
| Sample Volume | 5 mL |
| Sterility | Non-sterile |
| Filtration Area | 20 cm2 |
| Housing Material | Styrene acrylonitrile (SAN) |
| Quantity | 30/Pk. |
| Type | Static Utrafiltration Concentrator |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?