missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivacell™ 70 PES Centrifugal Concentrators

Product Code. 10410093 Shop All Sartorius Products
Change view
Click to view available options
MWCO:
0.2 μm
10 kDa
100 kDa
30 kDa
5 kDa
50 kDa
Quantity:
10/Pk.
2/Pk.
Unit Size:
Pack of 10
Pack of 2
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Quantity unitSize
10410093 5 kDa 2/Pk. Pack of 2
10283521 10 kDa 2/Pk. Pack of 2
10759571 10 kDa 10/Pk. Pack of 10
10079470 5 kDa 10/Pk. Pack of 10
10273761 30 kDa 2/Pk. Pack of 2
10739191 30 kDa 10/Pk. Pack of 10
10100711 50 kDa 2/Pk. Pack of 2
10769571 100 kDa 2/Pk. Pack of 2
10602622 50 kDa 10/Pk. Pack of 10
10479702 100 kDa 10/Pk. Pack of 10
10469512 0.2 μm 10/Pk. Pack of 10
10120711 0.2 μm 2/Pk. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 10410093 Supplier Sartorius Supplier No. VS6011

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Combines the ease of centrifugal devices with the control provided by pressure filtration. Sartorius Vivacell 70 PES Centrifugal Concentrators are disposable ultrafiltration units with the highest process flexibility for concentration of samples between 10 and 70ml. Options available for a wide range of MWCO.

Designed for ultra-fast concentrations
  • Features a vertical membrane design that minimizes fouling
  • Built-in deadstop of 150μl will prevent concentration to dryness
  • High flux PES membrane
  • High retentate recovery > 90%
Easy to handle
  • Disposable, easily assembled ultrafiltration unit
  • Requires no tubing connections or stirring mechanisms
  • Can be adapted to equipment availability or to specific user preferences
  • Universal rotor compatibility
Total process flexibility: centrifuge or pressure filtration
  • For convenience, the units are simply spun in a large capacity centrifuge (carriers accepting 250ml bottles)
  • For faster concentration, the device can be pressurized with air or inert gas before centrifuging
  • For better process control, gas pressure can be combined with gentle orbital shaking
  • For highest simplicity, the device is pressurized and can be left standing on a bench top or in a refrigerator
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material (Membrane) PES
MWCO 5 kDa
Housing Material Polycarbonate
Max. Force 75 psi
Quantity 2/Pk.
Volume (Metric) 10 to 70 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.