missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivacell™ 70 PES Centrifugal Concentrators
Shop All Sartorius Products
Click to view available options
MWCO:
0.2 μm
10 kDa
100 kDa
30 kDa
5 kDa
50 kDa
Quantity:
10/Pk.
2/Pk.
Unit Size:
Pack of 10
Pack of 2
Description
Designed for ultra-fast concentrations
- Features a vertical membrane design that minimizes fouling
- Built-in deadstop of 150μl will prevent concentration to dryness
- High flux PES membrane
- High retentate recovery > 90%
- Disposable, easily assembled ultrafiltration unit
- Requires no tubing connections or stirring mechanisms
- Can be adapted to equipment availability or to specific user preferences
- Universal rotor compatibility
- For convenience, the units are simply spun in a large capacity centrifuge (carriers accepting 250ml bottles)
- For faster concentration, the device can be pressurized with air or inert gas before centrifuging
- For better process control, gas pressure can be combined with gentle orbital shaking
- For highest simplicity, the device is pressurized and can be left standing on a bench top or in a refrigerator
Specifications
Specifications
| Disposable | Single-use |
| Material (Membrane) | PES |
| MWCO | 30 kDa |
| Housing Material | Polycarbonate |
| Max. Force | 75 psi |
| Quantity | 2/Pk. |
| Volume (Metric) | 10 to 70 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction