missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Sterility Testing Reusable System: Closing Ring

Product Code. 15982640 Shop All Sartorius Products
Click to view available options
:
Closing Ring, For filter holder 16522 (85mL)
This item is not returnable. View return policy

Product Code. 15982640

Brand: Sartorius™ 6980590

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Sterility testing System

Closing Ring - 14-555-945

  • Spare part for reusable sterility test system
  • For filter holder 16522 (85mL)

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Filter Holder (85 mL)
Type Closing Ring
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.