missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Stainless-steel Syringe Filter Holder

Product Code. 10424451 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric) Inside:
25 mm
47 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside unitSize
10424451 25 mm Each
10019450 47 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10424451 Supplier Sartorius Supplier No. 16214

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
  • Syringe Filter Holder is recommended for the filtration of volumes of up to about 100 ml.
  • In-Line Filter Holder has been designed for in-Line filtration with a pressure of up to 20 bar using 47mm Filter.
TRUSTED_SUSTAINABILITY

Specifications

Connectors Female luer lock (inlet), Male luer slip (outlet)
Disposable Reusable
Filter Type Manifold
Material PTFE-coated Stainless Steel
Sterility Non-sterile
Filtration Area 3 cm2
Max. Pressure 7 bar
Type Reusable Syringe Filter Holder
Volume (Metric) <0.1 mL
Diameter (Metric) Inside 25 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.