missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartolab™ RF/BT Vacuum Filtration Units

Product Code. 11711015 Shop All Sartorius Products
Change view
Click to view available options
Type:
Sartolab BT
Sartolab RF Filter
Filtration Area:
18 cm2
24 cm2
39 cm2
62 cm2
Volume (Metric):
150 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Pack of 48
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Filtration Area Volume (Metric) unitSize
11711015 Sartolab BT 62 cm2 1000 mL Pack of 12
11771005 Sartolab RF Filter 18 cm2 150 mL Pack of 12
11781005 Sartolab RF Filter 39 cm2 500 mL Pack of 12
11755815 Sartolab BT 18 cm2 150 mL Pack of 48
11721015 Sartolab RF Filter 24 cm2 250 mL Pack of 12
11701015 Sartolab BT 39 cm2 500 mL Pack of 12
11791005 Sartolab RF Filter 62 cm2 1000 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11711015 Supplier Sartorius Supplier No. 180C6E

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Filter aqueous solutions quickly and easily. Sartorius Sartolab™ RF/BT Vacuum Filtration Units are disposable and easy to use filter units. They contain a high quality PES membrane and combine the highest flow-rates and throughput with extremely low protein binding and extractables. Available in a range of volumes.

Quality, reliable sterile filtration units
  • Receiver Flask (RF) and Bottle Top (BT) units contain a high quality Sartorius PES membrane
  • Combine the highest flow-rates and throughput with extremely low protein binding and extractables
  • Ideal for filtration of tissue culture media, buffers, additives and other aqueous biologicals
  • RF and BT vacuum units are available as 150mL, 250mL, 500mL and 1000mL units
    • 500mL and 1000mL units come standard with 90mm membranes
  • Improved filtration speed and throughput for cost effective filtration
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP Class VI, ISO 10993
Connectors Plug-on hose nipple
Disposable Single-use
For Use With (Equipment) 1.77 in. bottle neck
Material Polystyrene
Material (Membrane) PES
Pore Size 0.22 μm
Sterility Sterile
Filtration Area 62 cm2
For Use With (Application) Filtration of tissue culture media, buffers, additives and other aqueous biologicals
Quantity 12 Case
Type Sartolab BT
Volume (Metric) 1000 mL
Volume (Metric) Hold Up 1000 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.