missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Quality, reliable sterile filtration units
- Receiver Flask (RF) and Bottle Top (BT) units contain a high quality Sartorius PES membrane
- Combine the highest flow-rates and throughput with extremely low protein binding and extractables
- Ideal for filtration of tissue culture media, buffers, additives and other aqueous biologicals
- RF and BT vacuum units are available as 150mL, 250mL, 500mL and 1000mL units
- 500mL and 1000mL units come standard with 90mm membranes
- Improved filtration speed and throughput for cost effective filtration
Specifications
Specifications
| Certifications/Compliance | USP Class VI, ISO 10993 |
| Connectors | Plug-on hose nipple |
| Disposable | Single-use |
| For Use With (Equipment) | 1.77 in. bottle neck |
| Material | Polystyrene |
| Material (Membrane) | PES |
| Pore Size | 0.22 μm |
| Sterility | Sterile |
| Filtration Area | 39 cm2 |
| For Use With (Application) | Filtration of tissue culture media, buffers, additives and other aqueous biologicals |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?