Learn More
Sartorius Quantitative Grade 1289 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Marque: Sartorius FT-3-208-070
Code : NEW
Informations supplémentaires : Weight : 0.10000kg
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.Spécification
Disc | |
100 sec. | |
100/Pk. | |
70 mm |
3 to 5 μm | |
1290 | |
0.21 mm |