Learn More
Sartorius Printer Paper, Labels, and Ink for YDP Printers
Printer Paper, Labels, and Ink for use with YDP30 and YDP40 laboratory printers. Designed to produce quick, clear printouts. Choose regular or self-adhesive paper rolls with ink ribbons for thermal transfer printing or regular paper rolls only for direct thermal printing.
Brand: Sartorius 69Y03287
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Printer Paper, Labels, and Ink for use with YDP30 and YDP40 laboratory printers. Designed to produce quick, clear printouts. Choose regular or self-adhesive paper rolls with ink ribbons for thermal transfer printing or regular paper rolls only for direct thermal printing.
For use with YDP30 and YDP40 laboratory printers- YDP30 Premium Printer prints using both direct thermal and thermal transfer printing methods
- YDP40 Standard Printer utilizes direct thermal printing only
- Direct thermal printing is ideal for less stringent requirements (paper roll only)
- Thermal transfer printing creates fade-resistant printouts for use in regulated areas
- Option include both paper and ink ribbon
- Choose between standard or self-adhesive paper rolls
Specifications
White | |
24 m | |
280 m | |
Standard Printer Paper |
78.7 ft. | |
918.6 ft. | |
Standard Lab Printer YDP40 or Premium Lab Printer YDP40 | |
5 Pk. |