missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Polycarbonate Track Etch Membrane Filters

Manufactured from high grade polycarbonate film using track-etch technology.

Brand:  Sartorius 23006--47------N

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10347451

  • £160.00 / Pack of 100
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

Polycarbonate Track Etch Membrane retain particles on their surfaces. Their capillary pore structure is uniform and precise, with a narrow pore size distribution. Track-etch membranes are an excellent choice for accurate fractionation of particles because of their smooth, flat surface results in high particulate visibility. Track-etch technology offers the user distinct performance advantages when excellent surface capture and high sample visibility are required. Applications: Particulate analysis, epifluorescence miroscopy, fluid clarification, cytology, cell biology, bioassays, water microbiology, environmental analysis.

Specifications

Specifications

Autoclavable
0.5 μm
100/Pk.
47 mm
Polycarbonate Track-Etched (PCTE)
Particulate analysis; epifluorescence miroscopy; fluid clarification; cytology; cell biology; bioassays; water microbiology; environmental analysis.
23 to 27 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers