Learn More
Sartorius Entris™ Toploading Balances
When you need to get a heavy workload of repetitive applications done fast. Innocal Calibration Services avaliable on some models.
Brand:  Sartorius ENTRIS5201I-1S
Code : NEW
Additional Details : Weight : 3.00000kg
 View more versions of this product
Product Code. 15234946
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
  • New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Innocal Calibration Services
Available in items containing "i-1SUS" in Manufacturer's No.: 14-559-023, -024,- 025, -026, -027, -028, -029, -030.
Specifications
Specifications
RS-232C, 25-pin
100 mg
300 mg
100 mg
Toploading Balance
180 mm
7.7 lb.
5.2 kg
Internal Calibration
ENTRIS5201i-1S
1.5 sec.
303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.)
180 mm
3.5 kg