Learn More
Sartorius Interface Module for Cubis Balances
Interface Module for use with Cubis balances.
£194.00
Product Code | Brand | Connection Type | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connection Type | For Use With (Application) | Price | Quantity & Availability | ||||
15385601
|
Sartorius
YDO01MS-P |
Option P: RS-232 9 Pin, PS/2 | Connecting a PC or a keyboard |
£194.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Option R (YDO01MS-R): RS-232C data interface, 25-pin, for connection of Cubis accessory equipment
- Option B (YDO01MS-B): Bluetooth™ interface module for wireless connection of the YDP10BT data printer
- Option P (YDO01MS-P): RS-232C data interface, 9-pin, including PS/2 port for connecting a PC or a keyboard
Specifications
Cubis Balance | |
Data Interface |