Learn More
- Home
- Products
- Filters and Filtration
- Filtration Membrane Discs
- Sartorius Cellulose Acetate Membrane Filters
Sartorius Cellulose Acetate Membrane Filters
Product Code. 10746981 Shop All Sartorius Products| Product Code. | Pore Size | Diameter (Metric) | Quantity | unitSize |
|---|---|---|---|---|
| 10746981 | 0.2 μm | 47 mm | 100/Pk. | Pack of 100 |
| 10189130 | 0.8 μm | 47 mm | 100/Pk. | Pack of 100 |
| 10466551 | 0.8 μm | 13 mm | 100/Pk. | Pack of 100 |
| 10314021 | 0.8 μm | 50 mm | 100/Pk. | Pack of 100 |
| 10271601 | 0.8 μm | 25 mm | 100/Pk. | Pack of 100 |
| 15912450 | 0.8 μm | 90 mm | 100/Pk. | Pack of 100 |
| 10692392 | 0.65 μm | 47 mm | 100/Pk. | Pack of 100 |
| 10674532 | 0.65 μm | 50 mm | 100/Pk. | Pack of 100 |
| 10313401 | 0.45 μm | 100 mm | 25/Pk. | Pack of 25 |
| 10154221 | 0.45 μm | 30 mm | 100/Pk. | Pack of 100 |
| 10788701 | 0.45 μm | 50 mm | 100/Pk. | Pack of 100 |
| 10796881 | 0.45 μm | 13 mm | 100/Pk. | Pack of 100 |
| 10475021 | 0.45 μm | 25 mm | 100/Pk. | Pack of 100 |
| 10739091 | 0.45 μm | 90 mm | 25/Pk. | Pack of 25 |
| 12942661 | 0.45 μm | 85 mm | 100/Pk. | Pack of 100 |
| 10788511 | 0.45 μm | 47 mm | 100/Pk. | Pack of 100 |
| 12932661 | 0.45 μm | 47 mm | 100/Pk. | Pack of 100 |
| 10422951 | 0.2 μm | 13 mm | 100/Pk. | Pack of 100 |
| 10709671 | 0.2 μm | 25 mm | 100/Pk. | Pack of 100 |
| 10622042 | 0.2 μm | 30 mm | 100/Pk. | Pack of 100 |
| 10612561 | 0.22 μm | 50 mm | 100/Pk. | Pack of 100 |
| 10528001 | 0.22 μm | 90 mm | 25/Pk. | Pack of 25 |
| 10255511 | 1.2 μm | 100 mm | 25/Pk. | Pack of 25 |
| 10715902 | 1.2 μm | 25 mm | 100/Pk. | Pack of 100 |
| 10281981 | 1.2 μm | 47 mm | 100/Pk. | Pack of 100 |
| 10620183 | 5.0 μm | 47 mm | 100/Pk. | Pack of 100 |
| 10173701 | 5.0 μm | 25 mm | 100/Pk. | Pack of 100 |
| 10707751 | 0.65 μm | 25 mm | 100/Pk. | Pack of 100 |
Description
Combines high flow rates and thermal stability with very low adsorption characteristics. Sartorius Cellulose Acetate Membrane Filters are excellently suited for use in pressure filtration devices. Non-sterile filters in a variety of diameters and pore sizes are bulk-packed to save money and space.
- Ideal for colony counting, particle testing, microscopy and sterility testing
- High-quality CA membrane filters for use in pressure filtration devices
- Available in pore sizes ranging from 0.2μm up to 5.0μm
- 0.2μm filters are the standard for sterile filtration of aqueous solutions, such as nutrient media, buffers and sera
- 5μm filters can be used as prefilters to retain coarse particles, while allowing microorganisms through
- White, non-sterile filters without grids in a variety of diameters (13 to 100mm)
- Can be sterilized by autoclaving at 121°C or 134°C; gamma irradiated to 25 kGy, dry heat or ethvlene oxide
The results of publications on adsorption are difficult to correlate, as mostly different test substances, conditions and detection methods were used, and the membranes were tested without previously being sterilized.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | White |
| Disposable | Single-use |
| Filter Type | Membrane Filter |
| Format | Disc |
| Material | Cellulose Acetate |
| Material (Membrane) | Cellulose Acetate |
| Pore Size | 0.2 μm |
| Sterility | Non-sterile |
| For Use With (Application) | For use in pressure filtration devices; Sterile filtration of solution. |
| Hydrophobicity | Hydrophilic |
| Quantity | 100/Pk. |
| Thickness (Metric) | 120 μm |
| Type | Membrane Filter |
| Diameter (Metric) | 47 mm |
| Show More Show Less |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.