missing translation for 'onlineSavingsMsg'
Learn More

Sartorius arium™ Pro ASTM TYPE 1 - Ultrapure Water System, Benchtop

Produce ASTM type 1 water for critical analysis like HPLC, PCR, antibody production, cell culture media preparation or electrophoresis by adding a arium™ Pro ASTM TYPE 1 - Ultrapure Water System to your lab.

Brand:  Sartorius H2OPRO-DI-T


Code : NEW

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 11745006

  • £6370.00 / Each
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

The arium pro series consist of flexible systems with device configurations that can be specially custom-tailored to applications and feature an outstanding cost-benefit ratio. All systems meet and exceed ASTM Type 1 water quality and ensure the best, reproducible results. Per hour, an arium pro can deliver up to 120 l of ultrapure water with a conductivity of 0.055μS/cm (18.2 megaohm/cm at 25°C) and consistently high quality. When a Sartopore° 2 150 final filter is used, arium pro will deliver ultrapure water that is practically free of microorganisms.

 

  • Multiple configurations offered based on application needs
  • Bench integrated design
  • Optimized components and cartridge materials with topdown-flow technology
  • Optional, integrated UV lamp (185/254nm) for eliminating bacteria and organics; controlled by reliable TOC monitoring
  • Display with touch-activated functions and intuitive menu navigation
  • Integrated real time monitoring and data display: feed conductivity, total water consumption, pressure regulator
  • PIN-protected access to basic settings /service
  • Convenient SD card slot

 

 

  • System can be chosen based on daily work
  • Guarantees a long service life and optimal water purification
  • Reliable water quality ensures reproducible results
  • Easy operation; no user manual needed
  • Optimal control of constant water flow and consistent water quality without extra costs
  • Prevents user operating errors
  • Supports reliable recording of important data
Note: Filter is not included; sold separately.

 

Specifications

Specifications

ASTM
<1 CFU/L
Up to 2 L/min.
<5 product water, <50 feed water
35.6°F to 86°F
General
Ultrapure Water System (Filter not included; sold separately)
<1/mL
130
5°C to 45°C
2°C to 35°C
13.7 in.
19.37 in.
17.75 in.
59.5 to 63.9 lb.
100/240 V 50/60 Hz
50/60 Hz
18.2 Meg-ohm/cm
120 L of ultrapure water
Type 1
Arium Pro ASTM Type 1 Ultrapure DI Benchtop Water System
2°C to 30°C
80%
Benchtop
4 to 10
Adsorption by spherical activated Carbon, deionization, optional final particle and sterile filtration
35.6°F to 95°F
<1
35 cm
49.2 cm
45.1 cm
27 to 29 kg
100/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers