missing translation for 'onlineSavingsMsg'
Learn More

Sartorius arium™ advance EDI Electodeionization Water System, ASTM Type 2
GREENER_CHOICEBIOPHARMA

Product Code. 15809354 Shop All Sartorius Products
Change view
Click to view available options
Flow Rate:
10 L/hr.
5 L/hr.
Mounting:
Benchtop
Wall
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Mounting unitSize
15809354 5 L/hr. Wall Each
14831382 10 L/hr. Benchtop Each
14821382 10 L/hr. Wall Each
15307373 5 L/hr. Benchtop Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15809354 Supplier Sartorius Supplier No. H2OEDI1B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Outfit your lab with he Sartorisu™ arium™ advance EDI provide consistently high quality Type 2 water.

The arium™ advance EDI pure water system delivers Type II water with the best quality at rates of up to 10l/hr. The system attains the highest retention rates of ions along with optimal water yield, and reliably removes oxidants, heavy metal ions and particulates from feed water. The pure water produced is stored in arium™ bagtank systems. If a Sartopore™ 2 150 final filter is used in the system, the water is practically free of microorganisms.

  • Innovative iJust technology
  • Uses unique bagtank technology with dispense gun connections
  • User-friendly, easy operation
  • Unique pre-filter technology with activated carbon, phosphate-free catalyst and 5μm particle filter

Benefits:

  • Optimized water consumption
  • Meets customer's lab requirements and saves maintenance time
  • No user manual needed, intuitive operation of the menu
  • Highly efficient reduces colloids and chlorine, oxidizing agents, organic and water-soluble heavy metals

Applications:

  • Microbiological media and reagents
  • Solutions for chemical analysis and synthesis
  • Histology
  • ELISA, RIA
  • Buffers and pH solutions
  • Feed water for laboratory devices, such as autoclaves, glassware washers, humidifiers, water baths etc.
TRUSTED_SUSTAINABILITY

Specifications

Conductivity 0.2 to 0.07 Product water, Less than1500 feed water
Contaminant Rejection (Microbiological) Greater than99%
Contaminant Rejection (Organic) Greater than99%
Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate 5 L/hr.
Product Water Type Type 2
Remote Dispenser No
Resistivity <5 to 15 product water
TOC Monitor with UV Intensity Monitoring No
Total Organic Carbon <50 product water, <1000 feed water
Feedwater Temperature (English) 35.6°F to 86°F
Feedwater Temperature (Metric) 2°C to 30°C
For Use With (Application) Feed water for laboratory devices, such as autoclaves, Glassware washers, humidifiers, water baths etc
Humidity Range 80%
Mounting Wall
Particle Size <1/mL
pH Range 4 to 10
Storage Requirements 5°C to 45°C
Temperature Range (English) 35.6°F to 95°F
Temperature Range (Metric) 2°C to 35°C
Tolerance 4
Turbidity <1
Width (English) 13.7 in.
Width (Metric) 35 cm
Height (English) 19.7 in.
Height (Metric) 50.1 cm
Depth (English) 17.75 in.
Depth (Metric) 45.1 cm
Weight (English) 57.3 lb.
Weight (Metric) 26 kg
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz
Daily Output 1 to 10 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Type Lab Water Purification Systems - Type 2 (Pure)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.