Learn More
Sartorius Air Pressure Controller (APC) for Vivacell Models
Product Code. 10039780 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Vivacell 20, 70, 100 and 250
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10039780 Vivacell 20, 70, 100 and 250 Each
1 options
Product Code. 10039780 Supplier Sartorius Supplier No. VCA002
Only null left
Add to basket
Add to basket
Use in combination with Vivacell™ Products to regulate and apply pressure to Vivacell™ units. Can be used with any pressure source.
For Vivacell 20, 70, 100 and 250 models. Complete with pressure gauge, regulator, over-pressure safety valve, female connector to Sartorius Stedim Biotech pressure products and 1m extension line (4 mm pneumatic tubing) with male and female connectors and 1m of 6mm inlet tubing.
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Filter Type Ultrafiltration
For Use With (Equipment) Vivacell 20, 70, 100 and 250
Sterility Non-sterile
Type Air Pressure Controller