Learn More
Sartorius Accessories for Sterisart™ Sterility Testing Systems: Dummy Plug
Product Code. 10367551 Shop All Sartorius Products
Click to view available options
:
Dummy Plug; Sterile: No
Product Code. 10367551
Brand: Sartorius 17012E
Only null left
Add to basket
Add to basket
Luer lock Dummy Plug for use with Sterisart™ Sterility Testing Systems.
  • The Sterisart™ NF Sterility Testing System provides completely closed system sterility testing of pharmaceuticals, open containers, closed containers, pre-filled syringes, powders, and medical devices.
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
For Use With (Equipment) Sterisart Sterility Testing System
Sterility Non-sterile
Type Dummy Plug