Learn More
Sartorius Accessories for Sterisart™ Sterility Testing Systems: Cover
Product Code. 11700136 Shop All Sartorius Products
Change view
Click to view available options
Type:
Millipore Drainage Cover
Sterisart Sterility Test Unit Drainage Cover
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11700136 Millipore Drainage Cover Each
11790126 Sterisart Sterility Test Unit Drainage Cover Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11700136 Supplier Sartorius Supplier No. 1ZG0012
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Cover is an Accessory for Sterisart™ Sterility Testing Systems.
  • Covers for various accessories used with Sterisart Sterility Testing Systems
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
For Use With (Equipment) Millipore Drainage Container
Material Stainless Steel
Sterility Non-sterile
Type Millipore Drainage Cover