Learn More
Sartorius Accessories for Combisart™ Systems: Forceps
Product Code. 10787431 Shop All Sartorius Products
Click to view available options
Display Type:
None
Unit Size:
Each
Product Code. 10787431
Brand: Sartorius 16625
Only null left
Add to basket
Add to basket
Forceps for use with Sartoris Combisart Systems
  • Stainless Steel
  • Blunt tip
  • Can be flamed, also autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tweezers
Display Type None
Material Stainless Steel
For Use With (Application) Firm hold of the membrane filter
For Use With (Equipment) Vacuum filter holders and manifold system
Includes Blunt-edged Tips