missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Accessories for Combisart™ Systems: Filtrate Flasks

Filtrate flasks for use with Sartorius Combisart Systems

Brand:  Sartorius 16606

Code : NEW

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11745555

  • £97.65 / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

Filtrate flasks for use with Sartorius Combisart Systems

  • Suction flask for individual filter holders
  • Vacuum-resistant borosilicate glass, 3.3 (Duran 50 glass)
  • Plastic hose nipple acc. to DIN 12476
  • Stopper/tube
  • Sterilizable

Specifications

Specifications

Suction Flask
Translucent
For collecting the filtrate
9mm Plastic safety hose nipple
1000mL
Borosilicate Glass
Individual vacuum filter holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers