missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Absorbent Pads

Product Code. 10708711 Shop All Sartorius Products
Click to view available options
Color:
Black
Dark Green
Gray
Green
White
Media Type:
Azide
Bismuth Sulfite
Caso
Cetrimide
Chapman
Chromocult
ECD
Endo
Glucose Tryptone
Jus de Tomate
Lysine
MacConkey
Orange Serum
R2A
Sabouraud Agar
Schaufus Pottinger
Standard
Standard TTC
TGE
Teepol
Tergitol TTC
VLB-S7-S
Wallerstein
Weman
Yeast Extract
m-FC
Pore Size:
0.45μm
0.65μm
0.8μm
1.2μm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10708711

Brand: Sartorius 1407050N

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sterile, dehydrated culture media which is individually inserted in a petri dish and sterilized

Sartorius Nutrient Pad Sets are used in the membrane filter methods. The membrane filter method allows large sample volumes with low microbial count to be tested. Inhibitors, such as essential oils or disinfectants, can be rinsed out after filtration, before incubation.Practical and easy to handle, Nutrient Pads reduce labor and simplify many microbiological testing procedures. Microorganisms require the appropriate culture media for optimal growth.Nutrient Pad Sets are sterile, dehydrated culture media. Once they are moistened with 3.0 - 3.5ml of sterile and demineralized (or destilled) water they are ready-to-use immediately. The level of moisture is optimal when an excess ring of water surrounding the NPS is visible.All Nutrient Pad Set types are supplied with the appropriate membrane filters, which are also individually, sterile packaged. The membrane filters tailored to meet the special requirements of microbial detection are available with 47 or 50mm diameter. The standard NPS box contains 100 sterile Nutrient Pads, each of which is individually inserted in a petri dish and sterilized. Ten each of these petri dishes are sealed in an aluminum bag. This special packaging in bags protects the sensitive formula constituents of the Nutrient Pad during transport and storage from fluctuations in humidity and temperature. As a result, it guarantees the high quality of NPS throughout their entire shelf life up to 24 months. More than 30 different Nutrient Pad Set types are available to meet the diverse objectives of microbiological analysis. Beyond the European drinking water directive, they comply with other international regulations and recommendations: international pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA, etc.

  • trouble-free storage up to 24 month
  • economical by time- and cost-saving
  • according to international standards and references
  • easy handling
  • validation guide available
  • consistent quality
  • highly versatile
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN and ISO standards
Color White
Form Dehydrated
Manufacturing Origin Germany
Media Type Schaufus Pottinger
Product Type Nutrient Pad
Sterility Sterile
Test Type Microbial Detection
For Use With (Application) Yeasts and molds
Grid Color Green
Packaging Type Individually Packaged
Pore Size 0.65μm
Diameter (English) Outside 1.96 in.
Diameter (Metric) Outside 5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.