missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Absorbent Pads

Product Code. 10092411 Shop All Sartorius Products
Change view
Click to view available options
Color:
Black
Dark Green
Gray
Green
White
Media Type:
Azide
Bismuth Sulfite
Caso
Cetrimide
Chapman
Chromocult
ECD
Endo
Glucose Tryptone
Jus de Tomate
Lysine
MacConkey
Orange Serum
R2A
Sabouraud Agar
Schaufus Pottinger
Standard
Standard TTC
TGE
Teepol
Tergitol TTC
VLB-S7-S
Wallerstein
Weman
Yeast Extract
m-FC
Pore Size:
0.45μm
0.65μm
0.8μm
1.2μm
Unit Size:
Pack of 100
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Media Type Pore Size unitSize
10092411 White m-FC 0.45μm Pack of 100
11700666 Dark Green TGE 0.45μm Pack of 100
11750676 Green Wallerstein 0.45μm Pack of 100
10108681 Dark Green Yeast Extract 0.45μm Pack of 100
10622602 Black Chromocult 0.45μm Pack of 100
10637461 Dark Green Azide 0.45μm Pack of 100
10424271 Green Tergitol TTC 0.45μm Pack of 100
10624371 Dark Green Caso 0.45μm Pack of 100
10144231 Green Schaufus Pottinger 1.2μm Pack of 100
10028050 Green Chapman 0.45μm Pack of 100
10129330 Green ECD 0.45μm Pack of 100
10357451 Green MacConkey 0.45μm Pack of 100
10557521 White Endo 0.45μm Pack of 100
10179180 Green Standard TTC 0.45μm Pack of 100
10767361 Green Bismuth Sulfite 0.45μm Pack of 100
10078020 Gray Bismuth Sulfite 0.65μm Pack of 100
10707941 White VLB-S7-S 0.45μm Pack of 100
10537331 Green Lysine 0.65μm Pack of 100
10189420 Gray Orange Serum 0.45μm Pack of 100
10738131 Green Standard 0.45μm Pack of 100
10245751 Gray Weman 0.45μm Pack of 100
10184271 White Glucose Tryptone 0.45μm Pack of 100
10409511 White Teepol 0.45μm Pack of 100
10261841 Gray Sabouraud Agar 0.65μm Pack of 100
10708711 White Schaufus Pottinger 0.65μm Pack of 100
10748131 White Cetrimide 0.45μm Pack of 100
10595701 Green Jus de Tomate 0.45μm Pack of 100
11770666 Gray Schaufus Pottinger 0.65μm Pack of 100
11843282 Gray Schaufus Pottinger 0.8μm Pack of 100
10225611 Green R2A 0.45μm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
This item is not returnable. View return policy
Product Code. 10092411 Supplier Sartorius Supplier No. 1406850N

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sterile, dehydrated culture media which is individually inserted in a petri dish and sterilized

Sartorius Nutrient Pad Sets are used in the membrane filter methods. The membrane filter method allows large sample volumes with low microbial count to be tested. Inhibitors, such as essential oils or disinfectants, can be rinsed out after filtration, before incubation.Practical and easy to handle, Nutrient Pads reduce labor and simplify many microbiological testing procedures. Microorganisms require the appropriate culture media for optimal growth.Nutrient Pad Sets are sterile, dehydrated culture media. Once they are moistened with 3.0 - 3.5ml of sterile and demineralized (or destilled) water they are ready-to-use immediately. The level of moisture is optimal when an excess ring of water surrounding the NPS is visible.All Nutrient Pad Set types are supplied with the appropriate membrane filters, which are also individually, sterile packaged. The membrane filters tailored to meet the special requirements of microbial detection are available with 47 or 50mm diameter. The standard NPS box contains 100 sterile Nutrient Pads, each of which is individually inserted in a petri dish and sterilized. Ten each of these petri dishes are sealed in an aluminum bag. This special packaging in bags protects the sensitive formula constituents of the Nutrient Pad during transport and storage from fluctuations in humidity and temperature. As a result, it guarantees the high quality of NPS throughout their entire shelf life up to 24 months. More than 30 different Nutrient Pad Set types are available to meet the diverse objectives of microbiological analysis. Beyond the European drinking water directive, they comply with other international regulations and recommendations: international pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA, etc.

  • trouble-free storage up to 24 month
  • economical by time- and cost-saving
  • according to international standards and references
  • easy handling
  • validation guide available
  • consistent quality
  • highly versatile
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN and ISO standards
Color White
Form Dehydrated
Manufacturing Origin Germany
Media Type m-FC
Product Type Nutrient Pad
Sterility Sterile
Test Type Microbial Detection
For Use With (Application) E. coli and coliforms
Grid Color Green
Packaging Type Individually Packaged
Pore Size 0.45μm
Diameter (English) Outside 1.96 in.
Diameter (Metric) Outside 5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.