Learn More
Sartorius Qualitative Grade 1290 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Brand: Sartorius FT-3-208-110
This item is not returnable.
View return policy
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Disc | |
Cotton Linters, Refined Pulp | |
3 to 5 μm | |
1290 | |
0.21 mm | |
110 mm |
Conical Funnel | |
3 to 5 μm | |
100 sec. | |
100/Pk. | |
Qualitative Filter Paper |