missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartolab P20 Plus
BIOPHARMA

Product Code. 18508074 Shop All Sartorius Products
Change view
Click to view available options
Pore Size:
0.45 μm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
18508074 0.45 μm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18508074 Supplier Sartorius Supplier No. 18076N
  • ONLINE EXCLUSIVE PRICE
    £824.00
    £798.00 / Pack of 100
    Save £26.00
    3% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pressure filtration unit with prefilter for the clarification of particle-high-loaded solutions ranging from 100 ml to 5 L.

  • It features hose barb inlet and a hose barb outlet
  • Includes a 0.45 μm polyethersulfone (PES) membrane
  • Available in a package containing 100 non-sterile pieces
  • Recommended for difficult-to-filter solutions like media that contains serum
  • Highest flow rates with a large surface of filtration (20 cm²)
  • No loss of protein with a low binding membrane
  • Low dead volume due to an optimized membrane support
  • Versions available with a prefilter for high particle load solutions
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Prefilter
Material Polycarbonate
Material (Membrane) PES
Pore Size 0.45 μm
Sterility Non-sterile
Filtration Area 20 cm²
Type Filter units
Volume (Metric) 1.2 mL
Diameter (Metric) 61 mm
For Use With (Equipment) Sartolab P20
Sample Volume 10 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.