missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartolab™ BT

Product Code. 31891589 Shop All Sartorius Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
Unit Size:
Pack of 24
Sample Volume:
0.15 μL
0.25 μL
0.5 μL
1 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize Sample Volume
31891589 0.22 μm Pack of 24 -
31891586 0.22 μm Pack of 24 0.15 μL
31891591 0.22 μm Pack of 24 0.25 μL
31891585 0.22 μm Pack of 24 0.5 μL
31891588 0.45 μm Pack of 24 1 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31891589 Supplier Sartorius Supplier No. 180E152
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

BT bottle top filter, a single-use vacuum filtration funnel designed for sterile filtration of aqueous samples

  • The Sartolab™ BT bottle top filter is comprised of a graduated funnel with a polyethersulfone (PES) membrane
  • A vacuum adapter and a lid, a tube connector for vacuum connection (for stand-alone filtration)
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
For Use With (Equipment) Bottle
Material Polystyrene
Pore Size 0.22 μm
Sterility Sterile
Type Bottle Top Filter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.