missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartoguard PES Midicaps 0.2µm size 7 OO connectors, 4/pack

Brand:  Sartorius 5475307F7--OO--A

398.86 GBP valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24159" to get your promotional price.


Sartoguard PES Midicaps 0.2µm size 7 OO connectors, 4/pack

Product Code. 18840681

  • £419.85 / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.