missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Sartoclean™ CA 10 in. Filter Cartridge with 25 Adapter

Brand:  Sartorius™ 5622506A1

Additional Details : Weight : 0.10000kg

Product Code. 10128871

  • £336.02 / Each
Estimated Shipment: 08-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Autoclavable
Cellulose Acetate
Sterile
Broad range of prefiltration applications in the biopharmaceutical industry
1Pack
CA 10 in. Filter Cartridge
USP, ISO 9001
0.45 μm
0.74 m2
Polypropylene
1Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers