Learn More
Sartorius™ Sartobind™ Polypropylene Housing Stabilized Reinforced Cellulose Membrane Q SingleSep Nano Capsules
Shop All Sartorius ProductsSpecifications
Specifications
| Certifications/Compliance | USP Class VI |
| Material (Membrane) | Anion Exchanger Stabilized Reinforced Cellulose |
| Pore Size | 3 to 5 μm |
| Filtration Area | 36 cm2 |
| For Use With (Application) | For DNA, host cell proteins, endotoxins, viruses contaminant removal applications |
| Housing Material | Polypropylene |
| No. per Pack | 4Pack |
| Quantity | 4Pack |
| Type | Q SingleSep Nano Capsules |