missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SART3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18025-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an RNA-binding nuclear protein that is a tumor-rejection antigen. This antigen possesses tumor epitopes capable of inducing HLA-A24-restricted and tumor-specific cytotoxic T lymphocytes in cancer patients and may be useful for specific immunotherapy. This gene product is found to be an important cellular factor for HIV-1 gene expression and viral replication. It also associates transiently with U6 and U4/U6 snRNPs during the recycling phase of the spliceosome cycle. This encoded protein is thought to be involved in the regulation of mRNA splicing.Specifications
| SART3 | |
| Polyclonal | |
| Unconjugated | |
| SART3 | |
| DSAP1, hSART 3, KIAA0156, P100, p110, p110(nrb), RP11 13G14, SART 3, SART3, TIP110 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 304582, 53890, 9733 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15020, Q9JLI8 | |
| Sart3 | |
| SART3 Fusion Protein Ag12590 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction