missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SART3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528625
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, H1299, HeLa, HepG2, Raji. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is an RNA-binding nuclear protein that is a tumor-rejection antigen. This antigen possesses tumor epitopes capable of inducing HLA-A24-restricted and tumor-specific cytotoxic T lymphocytes in cancer patients and may be useful for specific immunotherapy. This gene product is found to be an important cellular factor for HIV-1 gene expression and viral replication. It also associates transiently with U6 and U4/U6 snRNPs during the recycling phase of the spliceosome cycle. This encoded protein is thought to be involved in the regulation of mRNA splicing.
Specifications
| SART3 | |
| Polyclonal | |
| Unconjugated | |
| SART3 | |
| AU045857; DSAP1; HIV-1 Tat-interacting protein of 110kDa; hSART-3; Kiaa0156; MGC138188; mKIAA0156; mSART-3; P100; p110; p110 nuclear RNA-binding protein; p110(nrb); RP11-13G14; Sart3; SART-3; spliceosome associated factor 3, U4/U6 recycling protein; squamous cell carcinoma antigen recognised by T cells 3; squamous cell carcinoma antigen recognized by T cells 3; squamous cell carcinoma antigen recognized by T-cells 3; Tat-interacting protein of 110 kDa; Tip110; Tumor-rejection antigen SART3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9733 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.52 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q15020 | |
| SART3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SART3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction