missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SART2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | SART2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261542
|
Novus Biologicals
NBP2-58454 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647268
|
Novus Biologicals
NBP2-58454-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SART2 Polyclonal specifically detects SART2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| SART2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 29940 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MYWMDDTGYSEKATFASVTYPRGYPYNGTNYVNVTMHLRSPITRAAYLFIGPSIDVQSFTVHGDSQQLDVFIATSKHAYATYLWTGEATGQSAFAQVIADRHKILFDRNSAIKSSIVPEVKDYAAIVEQNLQH | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Chondroitin-glucuronate 5-epimerase, dermatan sulfate epimerase, dermatan-sulfate epimerase, DSEPI, EC 5.1.3.19, SART-2, SART2DS epimerase, squamous cell carcinoma antigen recognized by T cells 2, Squamous cell carcinoma antigen recognized by T-cells 2 | |
| DSE | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title