missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SART2 Antibody (3G11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42959
This item is not returnable.
View return policy
Description
SART2 Monoclonal antibody specifically detects SART2 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| SART2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 29940 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 3G11 | |
| Western Blot 1:200 - 1:1000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| Chondroitin-glucuronate 5-epimerase, dermatan sulfate epimerase, dermatan-sulfate epimerase, DSEPI, EC 5.1.3.19, SART-2, SART2DS epimerase, squamous cell carcinoma antigen recognized by T cells 2, Squamous cell carcinoma antigen recognized by T-cells 2 | |
| dermatan sulfate epimerase - (Uniprot# Q9UL01) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction