missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SART1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14836PE
This item is not returnable.
View return policy
Description
SART1 Polyclonal antibody specifically detects SART1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ICC/IF (Negative)
Specifications
| SART1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative) | |
| Allergen Hom s 1, Ara1, HAF, HOMS1, hSART-1, hSnu66, hypoxia-associated factor, IgE autoantigen, MGC2038, SART-1, SART1(259) protein, SART1(800) protein, SART1259, small nuclear ribonucleoprotein 110kDa (U4/U6.U5), SNRNP110, Snu66, SNU66 homolog, squamous cell carcinoma antigen recognised by T cells, squamous cell carcinoma antigen recognized by T cells, Squamous cell carcinoma antigen recognized by T cells 1, U4/U6.U5 tri-snRNP-associated 110 kDa protein, U4/U6.U5 tri-snRNP-associated protein 1 | |
| A synthetic peptide made to a internal portion of the human SART1 protein (between residues 100-200) [UniProt O43290] | |
| 0.1 mL | |
| Hypoxia | |
| 9092 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction