Learn More
Invitrogen™ SARS/SARS-CoV-2 Nucleocapsid Monoclonal Antibody (B46F)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17404
Description
MA1-7404 is reactive with the NP of SARS-CoV-2 (COVID-19 virus) by ELISA. Negative for human coronaviruses 229E & OC43. ELISA dilution range suggested 1:20- 1:200. The MA1-7404 antibody reacts with SARS Coronavirus nucleoprotein in viral and infected tissue samples. MA1-7404 has been successfully used in ELISA, Western blot and Immunofluorescence applications. The MA1-7404 antibody was raised against a SARS virus nucleoprotein preparation.
SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.
Specifications
| SARS/SARS-CoV-2 Nucleocapsid | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| P0DTC9 | |
| N | |
| SARS nucleoprotein preparation. | |
| 100 μg | |
| Primary | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| B46F | |
| Unconjugated | |
| N | |
| COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N | |
| Mouse | |
| Protein A | |
| RUO | |
| 43740575 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.