missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sars Membrane Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41059
Additional Details : Weight : 0.00970kg
Description
Sars Membrane Protein Polyclonal specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for ELISA.Specifications
Sars Membrane Protein | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
E1 glycoprotein, Matrix glycoprotein, Membrane glycoprotein, SARS coronavirus M protein, SARS coronavirus Membrane protein, SARS CoV, SARS CoV M glycoprotein, SARS CoV Matrix protein, Sars M, Sars Matrix Protein, SARSCoV, SARS-CoV M protein, SARS-CoV Membrane protein, Severe acute respiratory syndrome | |
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to a 13 aa peptide from within the first 50 amino acids from the M (Membrane protein) for the human SARS coronavirus (Genbank accession no. AFR58746.1) Amino Acid Squence: ADNGTITVEELKQ | |
0.1 mg | |
Primary | |
SARS-CoV | |
IgG |
ELISA | |
1 mg/mL | |
ELISA 1 μg/mL | |
P59596 | |
Rabbit | |
Affinity Purified | |
RUO | |
1489672 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |