missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sars Membrane Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41060
Additional Details : Weight : 0.00970kg
Description
Sars Membrane Protein Polyclonal specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for Western Blot, ELISA.Specifications
Sars Membrane Protein | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
E1 glycoprotein, Matrix glycoprotein, Membrane glycoprotein, SARS coronavirus M protein, SARS coronavirus Membrane protein, SARS CoV, SARS CoV M glycoprotein, SARS CoV Matrix protein, Sars M, Sars Matrix Protein, SARSCoV, SARS-CoV M protein, SARS-CoV Membrane protein, Severe acute respiratory syndrome | |
Antibody was raised against a synthetic peptide corresponding to 15 amino acids at the carboxy terminus of the SARS matrix protein. The immunogen is located within the last 50 amino acids of SARS Matrix. Amino Acid Squence: GNYKLNTDHAGSNDN | |
0.1 mg | |
Primary | |
SARS-CoV | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1 μg/mL, ELISA 1 μg/mL | |
P59596 | |
Rabbit | |
Affinity Purified | |
RUO | |
1489672 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |