missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sars Membrane Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £422.00
Specifications
| Antigen | Sars Membrane Protein |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682946
|
Novus Biologicals
NBP2-41059-0.025mg |
0.025 mg |
£150.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18105019
|
Novus Biologicals
NBP2-41059 |
0.1 mg |
£422.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sars Membrane Protein Polyclonal specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for ELISA.Specifications
| Sars Membrane Protein | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| E1 glycoprotein, Matrix glycoprotein, Membrane glycoprotein, SARS coronavirus M protein, SARS coronavirus Membrane protein, SARS CoV, SARS CoV M glycoprotein, SARS CoV Matrix protein, Sars M, Sars Matrix Protein, SARSCoV, SARS-CoV M protein, SARS-CoV Membrane protein, Severe acute respiratory syndrome | |
| The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to a 13 aa peptide from within the first 50 amino acids from the M (Membrane protein) for the human SARS coronavirus (Genbank accession no. AFR58746.1) Amino Acid Squence: ADNGTITVEELKQ | |
| Primary |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| SARS-CoV | |
| P59596 | |
| 1489672 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title