missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sars Membrane Protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£167.11 - £310.97
Specifications
Antigen | Sars Membrane Protein |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence reported by customer review |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18033985
|
Novus Biologicals
NB100-56569SS |
0.025 mg |
£167.11
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18006964
|
Novus Biologicals
NB100-56569 |
0.1 mg |
£310.97
0.10mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sars Membrane Protein Polyclonal specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for Western Blot.Specifications
Sars Membrane Protein | |
Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence reported by customer review | |
Polyclonal | |
Purified | |
RUO | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
E1 glycoprotein, Matrix glycoprotein, Membrane glycoprotein, SARS coronavirus M protein, SARS coronavirus Membrane protein, SARS CoV, SARS CoV M glycoprotein, SARS CoV Matrix protein, Sars M, Sars Matrix Protein, SARSCoV, SARS-CoV M protein, SARS-CoV Membrane protein, Severe acute respiratory syndrome | |
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 195-210 (YSRYRIGNYKLNTDH) from the M (Membrane protein) for the Human SARS coronavirus (Genbank accession no. YP_009724393.1) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
SARS-CoV | |
P59596 | |
1489672 | |
IgG | |
Protein G purified | |
The NCBI accession number of the SARS matrix protein is NP_828855.1. |
For Research Use Only