missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sars Membrane Protein Antibody (2H2C4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28852-0.1ML
Additional Details : Weight : 0.00970kg
Description
Sars Membrane Protein Monoclonal specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for Western Blot, ELISA.Specifications
Sars M | |
Monoclonal | |
Unconjugated | |
Subclonal supernatant with No Preservative | |
Purified recombinant fragment of SARS-m protein expressed in E. Coli. | |
0.1 mL | |
Specific for SARS-M. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
2H2C4 | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Mouse | |
Tissue culture supernatant | |
Primary | |
Virus | |
Supernatant |