missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sars Membrane Protein Antibody (2H2C4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28852-0.025ml
This item is not returnable.
View return policy
Description
Sars Membrane Protein Monoclonal specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for Western Blot, ELISA.
Specifications
| Sars M | |
| Monoclonal | |
| Unconjugated | |
| Subclonal supernatant with No Preservative | |
| Purified recombinant fragment of SARS-m protein expressed in E. Coli. | |
| 0.025 mL | |
| Specific for SARS-M. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 2H2C4 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Mouse | |
| Tissue culture supernatant | |
| Primary | |
| Virus | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction