missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV/SARS-CoV-2 NP Chimeric Recombinant Human Monoclonal Antibody (CR3009 (03-009))
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551595
This item is not returnable.
View return policy
Description
This antibody recognizes and binds the non-linear/conformational epitope of the N protein of SARS CoV and also binds the SARS CoV2 nucleoprotein.
Human coronaviruses are the major cause of upper respiratory tract illness in humans, such as the common cold. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The most prominent protein in the culture supernatants infected with SARS virus is a 46 kDa nucleocapsid protein. This suggests that the nucleocapsid protein is a major immunogen that may be useful for early diagnostics. SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus.
Specifications
| SARS-CoV/SARS-CoV-2 NP Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Human | |
| Affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgM κ |
| ELISA, Immunocytochemistry | |
| CR3009 (03-009) | |
| Unconjugated | |
| nucleoprotein | |
| Whole irradiated virion. | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction