missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Spike S2 Antibody (AbF804) - Azide and BSA Free, Novus Biologicals™

Product Code. 18393085 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393085 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393085 Supplier Novus Biologicals Supplier No. NBP307957

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

SARS-CoV-2 Spike S2 Monoclonal specifically detects SARS-CoV-2 Spike S2 in SARS-CoV-2 samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Spike S2
Applications Western Blot, ELISA
Classification Monoclonal
Clone AbF804
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, ELISA 1:5000-1:10000
Formulation PBS, pH 7.5
Gene Alias COVID-19, S2 protein, SARS-CoV-2, SARS-CoV-2 Spike, Spike glycoprotein Subunit 2, spike protein, Spike protein S2, structural protein, surface glycoprotein
Host Species Human
Immunogen Recombinant SARS-CoV-2 S2 protein
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Target Species SARS-CoV-2
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.