Learn More
SARS-CoV-2 Spike S1 Protein Antibody (1035206) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB105403B
365.50 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18629343
  • £442.00 / 0.10mL
Description
SARS-CoV-2 Spike S1 Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SARS-CoV-2 Spike S1 Protein
Monoclonal
Biotin
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
SARS-CoV-2
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
1035206
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready
SARS-CoV-2
HEK293-derived SARS-CoV-2 Spike S1 Subunit protein, Accession # YP_009724390.1
0.1 mL
Primary
Store at 4°C in the dark.
IgG1