missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Spike S1 Antibody (H4) - Azide and BSA Free, Novus Biologicals™

Product Code. 18350224 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350224 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350224 Supplier Novus Biologicals Supplier No. NBP307032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

SARS-CoV-2 Spike S1 Monoclonal specifically detects SARS-CoV-2 Spike S1 in SARS-CoV-2 samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Spike S1
Applications Western Blot, ELISA
Classification Monoclonal
Clone B38
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:5000-1:10000
Formulation PBS, pH 7.5
Gene Alias SARS-CoV-2
Host Species Human
Immunogen Recombinant SARS-CoV-2 Spike S1 protein
Purification Method >95%
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Target Species SARS-CoV-2
Content And Storage Store at 4°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.