missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Spike RBD Antibody (T3P1-C8) - VHH - Azide and BSA Free, Novus Biologicals™

Product Code. 18341396 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341396 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341396 Supplier Novus Biologicals Supplier No. NBP312849

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Llama Monoclonal Antibody

SARS-CoV-2 Spike RBD Monoclonal antibody specifically detects SARS-CoV-2 Spike RBD in SARS-CoV-2 samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Spike RBD
Applications ELISA
Classification Monoclonal
Clone T3P1-C8
Conjugate Unconjugated
Formulation PBS
Gene Alias 2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein
Host Species Llama
Immunogen SARS-CoV-2 S protein RBD containing C-terminal His Tag. The protein was expressed in human 293 cells (HEK293). It contains amino acids Arg 319 - Lys 537.
Purification Method Affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Target Species SARS-CoV-2
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.