missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Spike RBD Antibody (RBD5324) - Chimeric, Novus Biologicals™

Product Code. 18369561
Change view
Click to view available options
Quantity:
0.2 mg
Unit Size:
0.2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369561 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369561 Supplier Bio-Techne Supplier No. NBP312088

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

SARS-CoV-2 Spike RBD Monoclonal antibody specifically detects SARS-CoV-2 Spike RBD in SARS-CoV-2 samples. It is validated for ELISA, ELISA, Lateral Flow
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Spike RBD
Applications ELISA, Lateral Flow
Classification Monoclonal
Clone RBD5324
Conjugate Unconjugated
Dilution ELISA, Sandwich ELISA, Lateral Flow Assay
Formulation PBS, pH 7.4
Gene Alias 2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein
Host Species Rat
Immunogen SARS-CoV-2 Spike RBD
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Reconstitution String.Empty
Target Species SARS-CoV-2
Content And Storage Store at 4°C. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.