missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike RBD Antibody, R&D Systems™

Mouse Monoclonal Antibody
£185.00 - £483.00
Specifications
| Antigen | Spike RBD |
|---|---|
| Clone | 1049621 |
| Dilution | Blockade of Receptor-ligand Interaction, Neutralization, Immunocytochemistry 8-25 ug/mL |
| Applications | Blocking Assay, Neutralization, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18733937
|
R&D Systems
MAB11292-SP |
25 μg |
£185.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18783657
![]() |
R&D Systems
MAB11292-100 |
100 μg |
£483.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Blocking Assay, Neutralization, ImmunocytochemistrySpecifications
| Spike RBD | |
| Blockade of Receptor-ligand Interaction, Neutralization, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Human embryonic kidney cell, HEK293-derived SARS-CoV2 SARS-CoV2 spike, Arg319-Phe541, Accession # P0DTC2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1049621 | |
| Blocking Assay, Neutralization, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| SARS-CoV-2 | |
| P0DTC2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title